Product Details

SNP ID
rs8014028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90869408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGAAACTGGTCTTTCTTAACA[A/T]CTTTCCTCTATTATCTTCCTTTTCT
Phenotype
MIM: 603607
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KA5 PubMed Links

Gene Details

Gene
RPS6KA5
Gene Name
ribosomal protein S6 kinase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322227.1 5182 UTR 3 NP_001309156.1
NM_001322228.1 5182 UTR 3 NP_001309157.1
NM_001322229.1 5182 UTR 3 NP_001309158.1
NM_001322230.1 5182 UTR 3 NP_001309159.1
NM_001322231.1 5182 UTR 3 NP_001309160.1
NM_001322232.1 5182 UTR 3 NP_001309161.1
NM_001322233.1 5182 UTR 3 NP_001309162.1
NM_001322234.1 5182 UTR 3 NP_001309163.1
NM_001322235.1 5182 UTR 3 NP_001309164.1
NM_001322236.1 5182 UTR 3 NP_001309165.1
NM_001322237.1 5182 UTR 3 NP_001309166.1
NM_001322238.1 5182 UTR 3 NP_001309167.1
NM_004755.3 5182 UTR 3 NP_004746.2
NM_182398.2 5182 Intron NP_872198.1
XM_017021786.1 5182 Intron XP_016877275.1

View Full Product Details