Product Details

SNP ID
rs4387697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7790577 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGGCATGGGGCTGTGGATAGAC[C/G]CACCACGTGATGAGAAGGCTGGCCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4GP1 PubMed Links

Gene Details

Gene
CLEC4GP1
Gene Name
C-type lectin domain family 4 member G pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details