Product Details

SNP ID
rs28705211
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:965125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCAGCAGTGTGGGTGTGGCGGT[C/G]CTGGAGCTGCTCAATTTCCCGCCGC
Phenotype
MIM: 610770
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLHL17 PubMed Links

Gene Details

Gene
KLHL17
Gene Name
kelch like family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198317.2 2523 Silent Mutation GTC,GTG V621V NP_938073.1
XM_006710600.3 2523 Silent Mutation GTC,GTG V644V XP_006710663.1
XM_006710601.3 2523 Intron XP_006710664.1
Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
There are no transcripts associated with this gene.

Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160184.1 2523 Intron NP_001153656.1
NM_032129.2 2523 Intron NP_115505.2
XM_006710944.3 2523 Intron XP_006711007.2
XM_011542248.2 2523 Intron XP_011540550.2
XM_017002474.1 2523 Intron XP_016857963.1
XM_017002475.1 2523 Intron XP_016857964.1
XM_017002476.1 2523 Intron XP_016857965.1
XM_017002477.1 2523 Intron XP_016857966.1
XM_017002478.1 2523 Intron XP_016857967.1
XM_017002479.1 2523 Intron XP_016857968.1

View Full Product Details