Product Details

SNP ID
rs1998405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:71292920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTTAGTATTATGTAAAATATT[C/G]TAAAATAAGTTTTATCATAGAATCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OGFRL1 PubMed Links

Gene Details

Gene
OGFRL1
Gene Name
opioid growth factor receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324266.1 Intron NP_001311195.1
NM_024576.4 Intron NP_078852.3
XM_011536117.2 Intron XP_011534419.1
XM_011536118.2 Intron XP_011534420.1
XM_017011303.1 Intron XP_016866792.1
XM_017011304.1 Intron XP_016866793.1
XM_017011305.1 Intron XP_016866794.1
XM_017011306.1 Intron XP_016866795.1

View Full Product Details