Product Details

SNP ID
rs1804644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44943576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCTTCAGGACCAAATAGCAGAG[C/T]GCTCTGAAGGGAAAGACCATTTCAG
Phenotype
MIM: 616049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PABPC1L PubMed Links

Gene Details

Gene
PABPC1L
Gene Name
poly(A) binding protein cytoplasmic 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001124756.2 775 Intron NP_001118228.1
Gene
TOMM34
Gene Name
translocase of outer mitochondrial membrane 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006809.4 775 Silent Mutation GCA,GCG A234A NP_006800.2
XM_005260254.2 775 Silent Mutation GCA,GCG A132A XP_005260311.1
XM_011528501.1 775 Silent Mutation GCA,GCG A208A XP_011526803.1
XM_017027600.1 775 Missense Mutation CAC,CGC H185R XP_016883089.1

View Full Product Details