Product Details

SNP ID
rs8067359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5505777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATGATTAGCTTTCCACAGTGG[G/T]TTTTCTCCCACTAGAGAGAACTTAG
Phenotype
MIM: 606636
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC728392 PubMed Links

Gene Details

Gene
LOC728392
Gene Name
uncharacterized LOC728392
There are no transcripts associated with this gene.

Gene
NLRP1
Gene Name
NLR family pyrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033053.2 Intron NP_001028225.1
NM_014922.4 Intron NP_055737.1
NM_033004.3 Intron NP_127497.1
NM_033006.3 Intron NP_127499.1
NM_033007.3 Intron NP_127500.1

View Full Product Details