Product Details

SNP ID
rs10108486
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:91003375 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAGAATAAGAGGACAGAATTAAAA[C/T]GTCAAGGGACATAATTGATAGGATA
Phenotype
MIM: 609864
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TMEM55A PubMed Links

Gene Details

Gene
TMEM55A
Gene Name
transmembrane protein 55A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018710.2 Intron NP_061180.1
XM_011517152.2 Intron XP_011515454.1

View Full Product Details