Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282145.1 | 3309 | Intron | NP_001269074.1 | ||
NM_001282146.1 | 3309 | UTR 3 | NP_001269075.1 | ||
NM_020742.3 | 3309 | Intron | NP_065793.1 | ||
NM_181332.2 | 3309 | Intron | NP_851849.1 | ||
XM_005274564.2 | 3309 | Intron | XP_005274621.1 | ||
XM_005274565.2 | 3309 | UTR 3 | XP_005274622.1 | ||
XM_005274566.4 | 3309 | Intron | XP_005274623.1 | ||
XM_006724504.3 | 3309 | UTR 3 | XP_006724567.1 | ||
XM_011545547.2 | 3309 | UTR 3 | XP_011543849.1 | ||
XM_011545548.2 | 3309 | Intron | XP_011543850.1 | ||
XM_017029690.1 | 3309 | UTR 3 | XP_016885179.1 | ||
XM_017029691.1 | 3309 | UTR 3 | XP_016885180.1 | ||
XM_017029692.1 | 3309 | Intron | XP_016885181.1 | ||
XM_017029693.1 | 3309 | UTR 3 | XP_016885182.1 |