Product Details

SNP ID
rs2285674
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:45264939 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGTCATTCCACGGGACTCAGAGAAT[A/C]CCACCCGGCTCAAGTGTAGGTCTGG
Phenotype
MIM: 604655
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K14 PubMed Links

Gene Details

Gene
MAP3K14
Gene Name
mitogen-activated protein kinase kinase kinase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003954.4 Intron NP_003945.2
XM_011525441.2 Intron XP_011523743.1
Gene
MAP3K14-AS1
Gene Name
MAP3K14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SPATA32
Gene Name
spermatogenesis associated 32
There are no transcripts associated with this gene.

View Full Product Details