Product Details

SNP ID
rs7349054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43043290 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGCGAGTCTGGCCCACTCCCTCA[A/G]GAAGTCCACAGGCTGGACGCGGCAG
Phenotype
MIM: 608955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506679 PubMed Links

Gene Details

Gene
LOC100506679
Gene Name
uncharacterized LOC100506679
There are no transcripts associated with this gene.

Gene
TTLL1
Gene Name
tubulin tyrosine ligase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012263.4 Intron NP_036395.1
XM_006724225.3 Intron XP_006724288.1
XM_011530108.2 Intron XP_011528410.1
XM_011530110.2 Intron XP_011528412.1
XM_017028751.1 Intron XP_016884240.1
XM_017028752.1 Intron XP_016884241.1
XM_017028753.1 Intron XP_016884242.1

View Full Product Details