Product Details

SNP ID
hCV11926450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113105851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGAGATTTCATCATGGTCTCC[C/T]AGGCCCTCAGGCTCCTCTGCCTTCT
Phenotype
MIM: 613878 MIM: 609499
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F7 PubMed Links

Gene Details

Gene
F7
Gene Name
coagulation factor VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000131.4 64 Nonsense Mutation CAG,TAG Q4* NP_000122.1
NM_001267554.1 64 Nonsense Mutation CAG,TAG Q4* NP_001254483.1
NM_019616.3 64 Nonsense Mutation CAG,TAG Q4* NP_062562.1
Gene
MCF2L
Gene Name
MCF.2 cell line derived transforming sequence like
There are no transcripts associated with this gene.

View Full Product Details