Product Details

SNP ID
hCV11926459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113110806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAGTGCAAGGAGGAGCAGTGC[C/T]CCTTCGAGGAGGCCCGGGAGATCTT
Phenotype
MIM: 613878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F7 PubMed Links

Gene Details

Gene
F7
Gene Name
coagulation factor VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000131.4 235 Missense Mutation CCC,TCC P83S NP_000122.1
NM_001267554.1 235 Intron NP_001254483.1
NM_019616.3 235 Missense Mutation CCC,TCC P61S NP_062562.1

View Full Product Details