Product Details

SNP ID
rs1892667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30812787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCATGGAAAATTCAAAAGTAGC[G/T]CATGACAAGTGGGAGGTCAAATTTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP8-1 PubMed Links

Gene Details

Gene
KRTAP8-1
Gene Name
keratin associated protein 8-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175857.3 466 UTR 3 NP_787053.1

View Full Product Details