Product Details
- SNP ID
-
rs1063138
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75874279 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCCAGCTGTAGCTCTGTCTCCCA[A/G]AAGTGAGGTCTGCAGGGGAACAGGG
- Phenotype
-
MIM: 611041
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM47
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75830455,rs77043448] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRIM47
- Gene Name
- tripartite motif containing 47
- Gene
- TRIM65
- Gene Name
- tripartite motif containing 65
There are no transcripts associated with this gene.
View Full Product Details