Product Details

SNP ID
rs17463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14097296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAACAAGCAGGGCTCCCCAGGGA[A/G]TAGGATGGGTGAGCAGGGAACGGTC
Phenotype
MIM: 601639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKACA PubMed Links

Gene Details

Gene
PRKACA
Gene Name
protein kinase cAMP-activated catalytic subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304349.1 Intron NP_001291278.1
NM_002730.3 Intron NP_002721.1
NM_207518.2 Intron NP_997401.1
XM_017026948.1 Intron XP_016882437.1
Gene
SAMD1
Gene Name
sterile alpha motif domain containing 1
There are no transcripts associated with this gene.

View Full Product Details