Product Details

SNP ID
rs1997231
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:44617725 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGGTTGGGTAGGAGTAAATGGAGC[A/G]AAGCAGATCCCAGAACCAGAAGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF23 PubMed Links

Gene Details

Gene
IGSF23
Gene Name
immunoglobulin superfamily member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205280.1 216 Intron NP_001192209.1
XM_017026317.1 216 Intron XP_016881806.1
XM_017026318.1 216 Intron XP_016881807.1
XM_017026319.1 216 Intron XP_016881808.1
XM_017026320.1 216 Intron XP_016881809.1
XM_017026321.1 216 Intron XP_016881810.1
XM_017026322.1 216 UTR 5 XP_016881811.1
XM_017026323.1 216 Intron XP_016881812.1
XM_017026324.1 216 Intron XP_016881813.1
XM_017026325.1 216 Intron XP_016881814.1
XM_017026326.1 216 UTR 5 XP_016881815.1

View Full Product Details