Product Details

SNP ID
rs5810
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:27099728 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AACCACCCCTGCTCCAGGCAGAGAC[A/G]CTCAGAGATGCCCTCAGCCCGCAGC
Phenotype
MIM: 107310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC9A1 PubMed Links

Gene Details

Gene
SLC9A1
Gene Name
solute carrier family 9 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003047.4 2901 UTR 3 NP_003038.2
XM_011542021.2 2901 UTR 3 XP_011540323.1

View Full Product Details