Product Details

SNP ID
rs1620229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158575469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATATGAGTTTATGTTTATTTC[A/G]GGTAATCTATTTGACCCACCCGTTT
Phenotype
MIM: 601402
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100996447 PubMed Links

Gene Details

Gene
LOC100996447
Gene Name
uncharacterized LOC100996447
There are no transcripts associated with this gene.

Gene
MLF1
Gene Name
myeloid leukemia factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130156.2 Intron NP_001123628.1
NM_001130157.2 Intron NP_001123629.1
NM_001195432.1 Intron NP_001182361.1
NM_001195433.1 Intron NP_001182362.1
NM_001195434.1 Intron NP_001182363.1
NM_022443.4 Intron NP_071888.1
XM_005247479.4 Intron XP_005247536.1
XM_005247480.2 Intron XP_005247537.1
XM_006713645.2 Intron XP_006713708.1
XM_011512852.2 Intron XP_011511154.1
XM_011512853.2 Intron XP_011511155.1
XM_011512854.2 Intron XP_011511156.1

View Full Product Details