Product Details
- SNP ID
-
rs1926313
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
35
- Location
-
Chr.1:64755307 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CTAAACTTGTCTTCTAGGTATTTGG[A/T]GCTTAATTTTATGTTTATTTACTAC
- Phenotype
-
MIM: 609953
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RAVER2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79848041] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RAVER2
- Gene Name
- ribonucleoprotein, PTB binding 2
View Full Product Details