Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001288968.1 | 2117 | Intron | NP_001275897.1 | ||
NM_001288969.1 | 2117 | Intron | NP_001275898.1 | ||
NM_001288970.1 | 2117 | Intron | NP_001275899.1 | ||
NM_001288971.1 | 2117 | Intron | NP_001275900.1 | ||
NM_001288972.1 | 2117 | Intron | NP_001275901.1 | ||
NM_025149.5 | 2117 | Intron | NP_079425.3 | ||
XM_006722110.3 | 2117 | Intron | XP_006722173.1 | ||
XM_011525294.1 | 2117 | Intron | XP_011523596.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267.2 | 2117 | Missense Mutation | ACC,ATC | T350I | NP_001258.2 |
XM_011524214.2 | 2117 | Missense Mutation | ACC,ATC | T431I | XP_011522516.1 |