Product Details

SNP ID
rs2068178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169767851 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATCAGGTAGTTGGTAAACTTTCT[C/T]TAGACAATCGGGTTTCTTGGCCATT
Phenotype
MIM: 608534 MIM: 606042 MIM: 602322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTRT3 PubMed Links

Gene Details

Gene
ACTRT3
Gene Name
actin related protein T3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032487.4 1075 Missense Mutation AAG,GAG K234E NP_115876.3
Gene
MYNN
Gene Name
myoneurin
There are no transcripts associated with this gene.

Gene
TERC
Gene Name
telomerase RNA component
There are no transcripts associated with this gene.

View Full Product Details