Product Details

SNP ID
rs2241617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87407442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATGAGTTTTTATGTTAAACACT[T/C]CCAACTGTCAGTATTACCATGAAAT
Phenotype
MIM: 609604
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3B PubMed Links

Gene Details

Gene
MAP1LC3B
Gene Name
microtubule associated protein 1 light chain 3 beta
There are no transcripts associated with this gene.

Gene
ZCCHC14
Gene Name
zinc finger CCHC-type containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015144.2 5715 UTR 3 NP_055959.1
XM_005255858.3 5715 Intron XP_005255915.2
XM_011522964.2 5715 Intron XP_011521266.1
XM_017023082.1 5715 Intron XP_016878571.1

View Full Product Details