Product Details

SNP ID
rs2269370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153930976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGAAGGCTCCATCCTGAGACAA[A/C]AAGTCCAGTGTGACCTGCCCTCTGT
Phenotype
MIM: 300023 MIM: 300013 MIM: 312420
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
There are no transcripts associated with this gene.

Gene
NAA10
Gene Name
N(alpha)-acetyltransferase 10, NatA catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256119.1 Intron NP_001243048.1
NM_001256120.1 Intron NP_001243049.1
NM_003491.3 Intron NP_003482.1
Gene
RENBP
Gene Name
renin binding protein
There are no transcripts associated with this gene.

View Full Product Details