Product Details

SNP ID
rs2276900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127965423 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGTGTCCAAACTGCCGCGCCGCCC[C/T]GCTCCGGGTTTGTCTTCCTCCCTCC
Phenotype
MIM: 611124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD18 PubMed Links

Gene Details

Gene
ABHD18
Gene Name
abhydrolase domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319305.1 163 UTR 5 NP_001306234.1
NM_001319306.1 163 UTR 5 NP_001306235.1
NM_001319307.1 163 UTR 5 NP_001306236.1
XM_005263238.4 163 UTR 5 XP_005263295.1
XM_006714318.3 163 UTR 5 XP_006714381.1
XM_006714320.3 163 UTR 5 XP_006714383.1
XM_006714322.3 163 Intron XP_006714385.1
XM_011532272.2 163 UTR 5 XP_011530574.1
XM_011532273.2 163 UTR 5 XP_011530575.1
XM_011532274.2 163 UTR 5 XP_011530576.1
XM_011532276.1 163 UTR 5 XP_011530578.1
XM_011532278.2 163 UTR 5 XP_011530580.1
XM_011532279.2 163 Intron XP_011530581.1
XM_017008631.1 163 UTR 5 XP_016864120.1
XM_017008632.1 163 Intron XP_016864121.1
Gene
MFSD8
Gene Name
major facilitator superfamily domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152778.2 163 Intron NP_689991.1
XM_005262893.1 163 Intron XP_005262950.1
XM_005262896.1 163 Intron XP_005262953.1
XM_005262897.2 163 Intron XP_005262954.1
XM_005262898.2 163 Intron XP_005262955.1
XM_011531830.1 163 Intron XP_011530132.1
XM_011531831.1 163 Intron XP_011530133.1
XM_011531832.1 163 Intron XP_011530134.1
XM_017007989.1 163 Intron XP_016863478.1

View Full Product Details