Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319305.1 | Intron | NP_001306234.1 | |||
NM_001319306.1 | Intron | NP_001306235.1 | |||
NM_001319307.1 | Intron | NP_001306236.1 | |||
XM_005263238.4 | Intron | XP_005263295.1 | |||
XM_006714318.3 | Intron | XP_006714381.1 | |||
XM_006714320.3 | Intron | XP_006714383.1 | |||
XM_006714322.3 | Intron | XP_006714385.1 | |||
XM_011532272.2 | Intron | XP_011530574.1 | |||
XM_011532273.2 | Intron | XP_011530575.1 | |||
XM_011532274.2 | Intron | XP_011530576.1 | |||
XM_011532276.1 | Intron | XP_011530578.1 | |||
XM_011532278.2 | Intron | XP_011530580.1 | |||
XM_011532279.2 | Intron | XP_011530581.1 | |||
XM_017008631.1 | Intron | XP_016864120.1 | |||
XM_017008632.1 | Intron | XP_016864121.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152778.2 | Intron | NP_689991.1 | |||
XM_005262893.1 | Intron | XP_005262950.1 | |||
XM_005262896.1 | Intron | XP_005262953.1 | |||
XM_005262897.2 | Intron | XP_005262954.1 | |||
XM_005262898.2 | Intron | XP_005262955.1 | |||
XM_011531830.1 | Intron | XP_011530132.1 | |||
XM_011531831.1 | Intron | XP_011530133.1 | |||
XM_011531832.1 | Intron | XP_011530134.1 | |||
XM_017007989.1 | Intron | XP_016863478.1 |