Product Details

SNP ID
rs2701131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52033566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCACTTTCTCTTGGGGCCCTGA[C/T]GCCTTCCATGGCAACTAAGTAGAGC
Phenotype
MIM: 139139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NR4A1 PubMed Links

Gene Details

Gene
NR4A1
Gene Name
nuclear receptor subfamily 4 group A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202233.1 Intron NP_001189162.1
NM_001202234.1 Intron NP_001189163.1
NM_002135.4 Intron NP_002126.2
NM_173157.2 Intron NP_775180.1
XM_005268822.3 Intron XP_005268879.1
XM_005268824.3 Intron XP_005268881.1
XM_006719363.1 Intron XP_006719426.1
XM_006719364.3 Intron XP_006719427.1
XM_017019247.1 Intron XP_016874736.1
XM_017019248.1 Intron XP_016874737.1
XM_017019249.1 Intron XP_016874738.1

View Full Product Details