Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001206955.1 | 48 | Intron | NP_001193884.1 | ||
NM_001206956.1 | 48 | Intron | NP_001193885.1 | ||
NM_175607.2 | 48 | Intron | NP_783200.1 | ||
NM_175613.2 | 48 | Intron | NP_783302.1 | ||
XM_006713004.3 | 48 | Intron | XP_006713067.1 | ||
XM_011533425.2 | 48 | Intron | XP_011531727.1 | ||
XM_011533426.2 | 48 | Intron | XP_011531728.1 | ||
XM_011533427.2 | 48 | Intron | XP_011531729.1 | ||
XM_011533428.2 | 48 | Intron | XP_011531730.1 | ||
XM_011533429.2 | 48 | Intron | XP_011531731.1 | ||
XM_011533430.2 | 48 | Intron | XP_011531732.1 | ||
XM_011533431.2 | 48 | Intron | XP_011531733.1 | ||
XM_017005782.1 | 48 | Intron | XP_016861271.1 | ||
XM_017005783.1 | 48 | UTR 5 | XP_016861272.1 | ||
XM_017005784.1 | 48 | UTR 5 | XP_016861273.1 | ||
XM_017005785.1 | 48 | Intron | XP_016861274.1 | ||
XM_017005786.1 | 48 | Intron | XP_016861275.1 | ||
XM_017005787.1 | 48 | Intron | XP_016861276.1 | ||
XM_017005788.1 | 48 | Intron | XP_016861277.1 |