Product Details
- SNP ID
-
rs2227556
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:73911715 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCTGGGGTCCTCCGGATTCCATC[C/T]ACAGCAGGGCCAGACTCTCCCCAGG
- Phenotype
-
MIM: 191840
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C10orf55
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs371199984] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C10orf55
- Gene Name
- chromosome 10 open reading frame 55
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001791.2 |
767 |
Silent Mutation |
GTA,GTG |
V142V |
NP_001001791.2 |
- Gene
- PLAU
- Gene Name
- plasminogen activator, urokinase
View Full Product Details