Product Details

SNP ID
rs2484047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89415259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCGCCGCCTCCGGGAAACTGG[A/T]CAGCGCCTGCAGAGCCCCCAGGCGC
Phenotype
MIM: 147690 MIM: 616135
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IFIT1 PubMed Links

Gene Details

Gene
IFIT1
Gene Name
interferon induced protein with tetratricopeptide repeats 1
There are no transcripts associated with this gene.

Gene
IFIT5
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012420.2 Intron NP_036552.1

View Full Product Details