Product Details

SNP ID
rs2791195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70771644 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAATGGAGCGAATGGAATCCAA[C/T]CTCTCTTCTTTCAACATCCAAGTAA
Phenotype
MIM: 607506 MIM: 612640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS14 PubMed Links

Gene Details

Gene
ADAMTS14
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
There are no transcripts associated with this gene.

Gene
TBATA
Gene Name
thymus, brain and testes associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318241.1 1775 Intron NP_001305170.1
NM_001318242.1 1775 Intron NP_001305171.1
NM_001318243.1 1775 Intron NP_001305172.1
NM_152710.3 1775 Intron NP_689923.3
XM_017015845.1 1775 UTR 3 XP_016871334.1
XM_017015846.1 1775 Intron XP_016871335.1
XM_017015847.1 1775 Intron XP_016871336.1
XM_017015848.1 1775 Intron XP_016871337.1
XM_017015849.1 1775 Intron XP_016871338.1
XM_017015850.1 1775 Intron XP_016871339.1
XM_017015851.1 1775 Intron XP_016871340.1
XM_017015852.1 1775 Intron XP_016871341.1
XM_017015853.1 1775 Intron XP_016871342.1
XM_017015854.1 1775 Intron XP_016871343.1
XM_017015855.1 1775 Intron XP_016871344.1
XM_017015856.1 1775 Intron XP_016871345.1
XM_017015857.1 1775 Intron XP_016871346.1
XM_017015858.1 1775 Intron XP_016871347.1
XM_017015859.1 1775 Intron XP_016871348.1
XM_017015860.1 1775 Intron XP_016871349.1
XM_017015861.1 1775 Intron XP_016871350.1
XM_017015862.1 1775 Intron XP_016871351.1

View Full Product Details