Product Details

SNP ID
rs2904664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17165864 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGAGCTGAAACATTTAACCATG[C/T]GGGTATTGTGGAGAAATATATCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC2 PubMed Links

Gene Details

Gene
ZDHHC2
Gene Name
zinc finger DHHC-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016353.4 Intron NP_057437.1
XM_011544544.2 Intron XP_011542846.1
XM_011544545.2 Intron XP_011542847.1
XM_011544546.2 Intron XP_011542848.1
XM_011544547.2 Intron XP_011542849.1
XM_011544548.2 Intron XP_011542850.1
XM_011544549.2 Intron XP_011542851.1

View Full Product Details