Product Details

SNP ID
rs2297785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102919614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGGGGGCCAACACCATCTTCCT[C/T]ACCAAGTTTTTCATGATGATGACCT
Phenotype
MIM: 607803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNNM2 PubMed Links

Gene Details

Gene
CNNM2
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017649.4 1322 Silent Mutation CTC,CTT L378L NP_060119.3
NM_199076.2 1322 Silent Mutation CTC,CTT L378L NP_951058.1
NM_199077.2 1322 Silent Mutation CTC,CTT L378L NP_951059.1
XM_005269933.4 1322 Silent Mutation CTC,CTT L378L XP_005269990.1
XM_006717908.2 1322 Silent Mutation CTC,CTT L378L XP_006717971.1
XM_011539911.2 1322 Silent Mutation CTC,CTT L378L XP_011538213.1
Gene
LOC107984265
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details