Product Details
- SNP ID
-
rs2994051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:131433488 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGGCTGCCTGGGGCCCACTTGG[A/T]TGGCTGCACCTTTGTCTCCTGCTGC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRRC2B
PubMed Links
Gene Details
- Gene
- PRRC2B
- Gene Name
- proline rich coiled-coil 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013318.3 |
|
Intron |
|
|
NP_037450.2 |
View Full Product Details