Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077440.1 | 6972 | UTR 3 | NP_001070908.1 | ||
NM_001077441.1 | 6972 | UTR 3 | NP_001070909.1 | ||
NM_001301038.1 | 6972 | UTR 3 | NP_001287967.1 | ||
NM_014739.2 | 6972 | UTR 3 | NP_055554.1 | ||
XM_005267237.3 | 6972 | Intron | XP_005267294.1 | ||
XM_017011529.1 | 6972 | Intron | XP_016867018.1 | ||
XM_017011530.1 | 6972 | Intron | XP_016867019.1 | ||
XM_017011531.1 | 6972 | Intron | XP_016867020.1 | ||
XM_017011532.1 | 6972 | Intron | XP_016867021.1 |