Product Details

SNP ID
rs2295829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102922666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGTGGGGCAAAGTCTCCTGGT[C/G]GGGTGCAAGGAGCCGAGGCGAGATG
Phenotype
MIM: 605799 MIM: 614062
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AMN PubMed Links

Gene Details

Gene
AMN
Gene Name
amnion associated transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030943.3 11 UTR 5 NP_112205.2
XM_011537202.2 11 UTR 5 XP_011535504.1
XM_011537203.2 11 Intron XP_011535505.1
Gene
CDC42BPB
Gene Name
CDC42 binding protein kinase beta
There are no transcripts associated with this gene.

View Full Product Details