Product Details
- SNP ID
-
rs2255282
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:22025547 on Build GRCh38
- Set Membership
-
JSNP
Validated
- Context Sequence [VIC/FAM]
- GGCGCTCCAAAGCTCGGGGAGTGGT[G/T]GTCTCCCTTTCCTGGAGTCATTTGT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00339
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143282757] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00339
- Gene Name
- long intergenic non-protein coding RNA 339
There are no transcripts associated with this gene.
- Gene
- LOC101928043
- Gene Name
- uncharacterized LOC101928043
There are no transcripts associated with this gene.
View Full Product Details