Product Details

SNP ID
rs2664566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:48624601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTGACAGGCGTCCCCCCGCTT[C/T]CTCCCCAGCTGTGACCTTTCCCTGA
Phenotype
MIM: 606905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PREX1 PubMed Links

Gene Details

Gene
PREX1
Gene Name
phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020820.3 5189 UTR 3 NP_065871.2
XM_011528934.2 5189 UTR 3 XP_011527236.1

View Full Product Details