Product Details

SNP ID
rs2653601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8921395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTCAGCTTCCCCTGGCGGGGTC[C/T]GCGGTGGCTTCATGTGGGTTGCTTT
Phenotype
MIM: 609191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKIP1 PubMed Links

Gene Details

Gene
AKIP1
Gene Name
A-kinase interacting protein 1
There are no transcripts associated with this gene.

Gene
C11orf16
Gene Name
chromosome 11 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020643.2 1910 Missense Mutation CAG,CGG Q442R NP_065694.2
XM_017018013.1 1910 Missense Mutation CAG,CGG Q442R XP_016873502.1

View Full Product Details