Product Details
- SNP ID
-
rs2690046
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:25285838 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTGGAGAATTTTAAAACAAAT[C/T]CCAGGTATGTTATTGAAAGAGTATA
- Phenotype
-
MIM: 609593
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CARMIL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111984173] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CARMIL1
- Gene Name
- capping protein regulator and myosin 1 linker 1
- Gene
- LOC105375106
- Gene Name
- uncharacterized LOC105375106
There are no transcripts associated with this gene.
View Full Product Details