Product Details
- SNP ID
-
rs3822057
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:186266998 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGGATTGGGAAAGAAAATTCAAC[A/C]TTATAATGAGAACACTGTGAATGCT
- Phenotype
-
MIM: 264900
MIM: 229000
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
F11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35709976] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- F11
- Gene Name
- coagulation factor XI
- Gene
- KLKB1
- Gene Name
- kallikrein B1
There are no transcripts associated with this gene.
View Full Product Details