Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000765.4 | 1333 | Missense Mutation | ACA,AGA | T409R | NP_000756.3 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256497.2 | 1333 | Missense Mutation | ACA,AGA | T409R | NP_001243426.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_145115.2 | 1333 | Intron | NP_660090.2 | ||
XM_005250194.2 | 1333 | Intron | XP_005250251.1 | ||
XM_011515905.2 | 1333 | Intron | XP_011514207.1 | ||
XM_011515906.2 | 1333 | Intron | XP_011514208.1 | ||
XM_011515907.2 | 1333 | Intron | XP_011514209.1 | ||
XM_011515908.2 | 1333 | Intron | XP_011514210.1 | ||
XM_011515909.2 | 1333 | Intron | XP_011514211.1 | ||
XM_011515910.2 | 1333 | Intron | XP_011514212.1 | ||
XM_017011824.1 | 1333 | Intron | XP_016867313.1 | ||
XM_017011825.1 | 1333 | Intron | XP_016867314.1 | ||
XM_017011826.1 | 1333 | Intron | XP_016867315.1 | ||
XM_017011827.1 | 1333 | Intron | XP_016867316.1 |