Product Details

SNP ID
rs4987678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142872464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAGCTTCCGCTCCAGCATCAC[C/T]GTGGTGGCCACAATCTGCGTATGGG
Phenotype
MIM: 602757 MIM: 606680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHB6 PubMed Links

Gene Details

Gene
EPHB6
Gene Name
EPH receptor B6
There are no transcripts associated with this gene.

Gene
TRPV6
Gene Name
transient receptor potential cation channel subfamily V member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018646.5 2032 Silent Mutation ACA,ACG T641T NP_061116.5

View Full Product Details