Product Details

SNP ID
rs33926104
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:40991390 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCAGCGCCACCCTAACACCCATGAC[A/C]GCCTCCCACCAGGGCCCCGCCCTCT
Phenotype
MIM: 123930
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
CYP2B6*17A,c.85C>A CYP2B6*17A,g.85C>A CYP2B6*17B,c.85C>A CYP2B6*17B,g.85C>A
Literature Links
CYP2B6 PubMed Links

Gene Details

Gene
CYP2B6
Gene Name
cytochrome P450 family 2 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000767.4 188 Missense Mutation AGC,CGC S29R NP_000758.1
XM_005258569.4 188 Missense Mutation AGC,CGC S29R XP_005258626.1
XM_006723050.3 188 Missense Mutation AGC,CGC S29R XP_006723113.1
XM_011526546.2 188 Missense Mutation AGC,CGC S29R XP_011524848.1
XM_011526547.2 188 Intron XP_011524849.1
XM_011526548.2 188 Missense Mutation AGC,CGC S29R XP_011524850.1
XM_011526549.2 188 Intron XP_011524851.1
XM_011526550.2 188 Intron XP_011524852.1

View Full Product Details