Product Details

SNP ID
rs10258348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73444153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGAAAGAAAAACTATGGATTCA[G/T]CAGAGAACAACGGAAGGTGAAGGGA
Phenotype
MIM: 605681 MIM: 601766
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAZ1B PubMed Links

Gene Details

Gene
BAZ1B
Gene Name
bromodomain adjacent to zinc finger domain 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032408.3 Intron NP_115784.1
XM_017012773.1 Intron XP_016868262.1
Gene
FZD9
Gene Name
frizzled class receptor 9
There are no transcripts associated with this gene.

View Full Product Details