Product Details

SNP ID
rs990560942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39923011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCAACTTCCTCTGTTCATCCC[C/T]ACACCAGGAGCCACGGCAGGGTTAA
Phenotype
MIM: 611221 MIM: 610075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSDMB PubMed Links

Gene Details

Gene
GSDMB
Gene Name
gasdermin B
There are no transcripts associated with this gene.

Gene
LRRC3C
Gene Name
leucine rich repeat containing 3C
There are no transcripts associated with this gene.

Gene
ORMDL3
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320801.1 Intron NP_001307730.1
NM_001320802.1 Intron NP_001307731.1
NM_001320803.1 Intron NP_001307732.1
NM_139280.3 Intron NP_644809.1

View Full Product Details