Product Details

SNP ID
rs3830026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116664061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGGCAGCTTCCATCTAGGAGAG[A/G]AACCGTCCGTGTGAGTTGCAGGGAG
Phenotype
MIM: 610701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf82 PubMed Links

Gene Details

Gene
C10orf82
Gene Name
chromosome 10 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144661.2 629 UTR 3 NP_653262.1
XM_011539340.2 629 Intron XP_011537642.1
XM_011539341.1 629 Intron XP_011537643.1
XM_011539342.2 629 Intron XP_011537644.1
XM_011539343.2 629 Intron XP_011537645.1
XM_011539344.2 629 Intron XP_011537646.1
XM_011539345.2 629 Intron XP_011537647.1
XM_011539346.2 629 Intron XP_011537648.1
XM_011539347.2 629 UTR 3 XP_011537649.1
XM_017015753.1 629 Intron XP_016871242.1
XM_017015754.1 629 Intron XP_016871243.1
XM_017015755.1 629 Intron XP_016871244.1
Gene
HSPA12A
Gene Name
heat shock protein family A (Hsp70) member 12A
There are no transcripts associated with this gene.

Gene
LOC105378499
Gene Name
uncharacterized LOC105378499
There are no transcripts associated with this gene.

View Full Product Details