Product Details

SNP ID
rs13029406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38802932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCTCACCTGTAACAAAAAACCT[C/T]AGATGATGACAGTGATGTCACTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX57 PubMed Links

Gene Details

Gene
DHX57
Gene Name
DEAH-box helicase 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011533154.2 Intron XP_011531456.1
XM_011533155.2 Intron XP_011531457.1
XM_011533156.2 Intron XP_011531458.1
XM_011533157.1 Intron XP_011531459.1

View Full Product Details