Product Details

SNP ID
rs61747755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70196591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCCTGGGAGCCACATTGTCAGC[A/C]CCAGTAAGTGATTTTATGGCACTAC
Phenotype
MIM: 614843
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ODAM PubMed Links

Gene Details

Gene
ODAM
Gene Name
odontogenic, ameloblast asssociated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017855.3 65 Silent Mutation GCA,GCC A16A NP_060325.3
XM_005265698.3 65 Silent Mutation GCA,GCC A33A XP_005265755.1
XM_006714245.3 65 Silent Mutation GCA,GCC A33A XP_006714308.1
XM_017008331.1 65 Silent Mutation GCA,GCC A16A XP_016863820.1

View Full Product Details