Product Details
- SNP ID
-
rs17042066
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:16271715 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAGAGAACTGCAAGTGTCCTTC[A/G]ACGGGAGGTTTGTATCTTTGGGGTA
- Phenotype
-
MIM: 608959
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DPH3
PubMed Links
Gene Details
- Gene
- DPH3
- Gene Name
- diphthamide biosynthesis 3
There are no transcripts associated with this gene.
- Gene
- OXNAD1
- Gene Name
- oxidoreductase NAD binding domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138381.3 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
NP_612390.1 |
XM_005265559.3 |
676 |
Missense Mutation |
CAA,CGA |
Q132R |
XP_005265616.1 |
XM_006713405.1 |
676 |
Missense Mutation |
CAA,CGA |
Q77R |
XP_006713468.1 |
XM_006713406.2 |
676 |
UTR 5 |
|
|
XP_006713469.1 |
XM_011534230.2 |
676 |
Missense Mutation |
CAA,CGA |
Q132R |
XP_011532532.1 |
XM_011534232.2 |
676 |
Missense Mutation |
CAA,CGA |
Q132R |
XP_011532534.1 |
XM_011534233.2 |
676 |
Missense Mutation |
CAA,CGA |
Q132R |
XP_011532535.1 |
XM_011534234.2 |
676 |
Missense Mutation |
CAA,CGA |
Q132R |
XP_011532536.1 |
XM_011534235.2 |
676 |
Missense Mutation |
CAA,CGA |
Q132R |
XP_011532537.1 |
XM_017007485.1 |
676 |
Missense Mutation |
CAA,CGA |
Q77R |
XP_016862974.1 |
XM_017007486.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862975.1 |
XM_017007487.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862976.1 |
XM_017007488.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862977.1 |
XM_017007489.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862978.1 |
XM_017007490.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862979.1 |
XM_017007491.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862980.1 |
XM_017007492.1 |
676 |
Missense Mutation |
CAA,CGA |
Q59R |
XP_016862981.1 |
XM_017007493.1 |
676 |
UTR 5 |
|
|
XP_016862982.1 |
XM_017007494.1 |
676 |
UTR 5 |
|
|
XP_016862983.1 |
XM_017007495.1 |
676 |
UTR 5 |
|
|
XP_016862984.1 |
XM_017007496.1 |
676 |
UTR 5 |
|
|
XP_016862985.1 |
View Full Product Details