Product Details

SNP ID
rs116626237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48713836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACCTGACTGGAGAACCCTAGC[C/T]GGATTCAACCCACACCCTCTTACCT
Phenotype
MIM: 182100 MIM: 610349 MIM: 609623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUT2 PubMed Links

Gene Details

Gene
FUT2
Gene Name
fucosyltransferase 2
There are no transcripts associated with this gene.

Gene
LOC105447645
Gene Name
uncharacterized LOC105447645
There are no transcripts associated with this gene.

Gene
MAMSTR
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130915.1 Intron NP_001124387.1
NM_001297753.1 Intron NP_001284682.1
NM_182574.2 Intron NP_872380.1
XM_011526807.2 Intron XP_011525109.1
XM_011526808.2 Intron XP_011525110.1
XM_011526809.2 Intron XP_011525111.1
XM_017026640.1 Intron XP_016882129.1
XM_017026641.1 Intron XP_016882130.1
XM_017026642.1 Intron XP_016882131.1
XM_017026643.1 Intron XP_016882132.1
XM_017026644.1 Intron XP_016882133.1
XM_017026645.1 Intron XP_016882134.1
Gene
RASIP1
Gene Name
Ras interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details